The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281768.1 | NP_001268697.1 | 2 - 3 | 342 | 52 |
NM_021116.2 | NP_066939.1 | 1 - 2 | 656 | 52 | |
XM_005249584.3 | XP_005249641.1 | 1 - 2 | 654 | 52 | |
XM_005249585.2 | XP_005249642.1 | 1 - 2 | 654 | 52 | |
GenBank mRNA | AB209496.1 | - | 1 - 2 | 601 | 52 |
AF497515.1 | - | 1 - 2 | 660 | 52 |