Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN15
Entrez Gene ID
23555
Gene Name
tetraspanin 15
Gene Aliases
2700063A19Rik, NET-7, NET7, TM4SF15
Chromosome Location
Chr.10: 69451464 - 69549508 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012339.3 NP_036471.1 2 - 3 403 63
XM_017016010.1 XP_016871499.1 2 - 3 409 63
GenBank mRNA AF120266.1 - 2 - 3 394 63
BC003157.1 - 2 - 3 397 63
BC004161.2 - 2 - 3 397 63

View Full Product Details