The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000616.4 | NP_000607.1 | 6 - 7 | 1216 | 128 |
NM_001195014.2 | NP_001181943.1 | 4 - 5 | 1260 | 128 | |
NM_001195015.2 | NP_001181944.1 | 4 - 5 | 948 | 128 | |
NM_001195016.2 | NP_001181945.1 | 4 - 5 | 935 | 128 | |
NM_001195017.2 | NP_001181946.1 | 4 - 5 | 913 | 128 | |
XM_017020228.1 | XP_016875717.1 | 4 - 5 | 1041 | 128 | |
GenBank mRNA | AK300052.1 | - | 4 - 5 | 831 | 128 |
AK303968.1 | - | 3 - 4 | 995 | 128 | |
AK310677.1 | - | 3 - 4 | 960 | 128 | |
AK312828.1 | - | 6 - 7 | 1153 | 128 | |
AK315898.1 | - | 4 - 5 | 1196 | 128 | |
AK316461.1 | - | 4 - 5 | 879 | 128 | |
AK316462.1 | - | 4 - 5 | 885 | 128 | |
BC025782.1 | - | 6 - 7 | 1110 | 128 | |
BT019791.1 | - | 5 - 6 | 958 | 128 | |
BT019811.1 | - | 5 - 6 | 958 | 128 | |
M12807.1 | - | 5 - 6 | 1033 | 128 |