The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016565.2 | NP_057649.2 | 1 - 2 | 86 | 63 |
GenBank mRNA | AK000434.1 | - | 1 - 2 | 86 | 63 |
AL359618.1 | - | 1 - 2 | 66 | 63 | |
BC106036.1 | - | 1 - 2 | 102 | 63 |