The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000368.4 | NP_000359.1 | 3 - 4 | 341 | 67 |
NM_001162426.1 | NP_001155898.1 | 3 - 4 | 341 | 67 | |
NM_001162427.1 | NP_001155899.1 | 3 - 4 | 341 | 67 | |
XM_005272211.1 | XP_005272268.1 | 3 - 4 | 386 | 67 | |
XM_006717271.1 | XP_006717334.1 | 3 - 4 | 321 | 67 | |
XM_011518979.2 | XP_011517281.1 | 3 - 4 | 335 | 67 | |
XM_017015096.1 | XP_016870585.1 | 4 - 5 | 800 | 67 | |
XM_017015097.1 | XP_016870586.1 | 4 - 5 | 771 | 67 | |
XM_017015098.1 | XP_016870587.1 | 3 - 4 | 386 | 67 | |
GenBank mRNA | AB190910.1 | - | 3 - 4 | 303 | 67 |
AF013168.1 | - | 3 - 4 | 328 | 67 | |
AK303030.1 | - | 3 - 4 | 324 | 67 | |
AK308412.1 | - | 3 - 4 | 324 | 67 | |
BC047772.2 | - | 3 - 4 | 314 | 67 | |
BC070032.1 | - | 3 - 4 | 342 | 67 | |
BC108668.1 | - | 3 - 4 | 331 | 67 | |
BC121000.2 | - | 3 - 4 | 298 | 67 | |
BI463204.1 | - | 3 - 4 | 66 | ||
DA726552.1 | - | 3 - 4 | 342 | 67 | |
KJ534953.1 | - | 1 - 2 | 107 | 67 |