The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014899.3 | NP_055714.3 | 3 - 4 | 950 | 110 |
XM_011543279.2 | XP_011541581.1 | 3 - 4 | 1030 | 110 | |
XM_017009236.1 | XP_016864725.1 | 4 - 5 | 711 | 110 | |
XM_017009237.1 | XP_016864726.1 | 4 - 5 | 832 | 110 | |
GenBank mRNA | AB020685.1 | - | 3 - 4 | 748 | 110 |
AK023621.1 | - | 3 - 4 | 544 | 110 | |
AK290034.1 | - | 3 - 4 | 723 | 110 | |
BC020231.1 | - | 3 - 4 | 919 | 110 | |
BC041337.2 | - | 3 - 4 | 755 | 110 |