The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014899.3 | NP_055714.3 | 8 - 9 | 1820 | 75 |
XM_011543279.2 | XP_011541581.1 | 7 - 8 | 1534 | 75 | |
XM_017009236.1 | XP_016864725.1 | 9 - 10 | 1581 | 75 | |
XM_017009237.1 | XP_016864726.1 | 9 - 10 | 1702 | 75 | |
GenBank mRNA | AB020685.1 | - | 8 - 9 | 1618 | 75 |
AK290034.1 | - | 8 - 9 | 1593 | 75 | |
BC041337.2 | - | 8 - 9 | 1625 | 75 |