The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318835.1 | NP_001305764.1 | 3 - 4 | 480 | 104 |
NM_005794.3 | NP_005785.1 | 3 - 4 | 768 | 104 | |
NM_182908.4 | NP_878912.1 | 3 - 4 | 768 | 104 | |
XM_005267249.1 | XP_005267306.1 | 3 - 4 | 480 | 104 | |
XM_006720001.3 | XP_006720064.1 | 3 - 4 | 443 | 104 | |
XM_011536338.1 | XP_011534640.1 | 2 - 3 | 409 | 104 | |
XM_011536339.2 | XP_011534641.1 | 2 - 3 | 259 | 104 | |
GenBank mRNA | AB209653.1 | - | 2 - 3 | 1154 | 104 |
AK222857.1 | - | 3 - 4 | 768 | 104 | |
AK309945.1 | - | 3 - 4 | 769 | 104 | |
AK309975.1 | - | 3 - 4 | 480 | 104 | |
BC002786.2 | - | 3 - 4 | 759 | 104 | |
BC007339.2 | - | 2 - 3 | 294 | 104 | |
U31875.1 | - | 3 - 4 | 754 | 104 |