The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005794.3 | NP_005785.1 | 6 - 7 | 989 | 100 |
NM_182908.4 | NP_878912.1 | 6 - 7 | 989 | 100 | |
XM_005267249.1 | XP_005267306.1 | 6 - 7 | 701 | 100 | |
XM_006720001.3 | XP_006720064.1 | 6 - 7 | 664 | 100 | |
XM_011536338.1 | XP_011534640.1 | 5 - 6 | 630 | 100 | |
XM_011536339.2 | XP_011534641.1 | 5 - 6 | 480 | 100 | |
GenBank mRNA | AB209653.1 | - | 4 - 5 | 4631 | 100 |
AK092834.1 | - | 2 - 3 | 2400 | 100 | |
AK309945.1 | - | 6 - 7 | 2476 | 100 | |
BC002786.2 | - | 6 - 7 | 980 | 100 | |
BC007339.2 | - | 5 - 6 | 515 | 100 | |
U31875.1 | - | 6 - 7 | 975 | 100 |