The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020795.3 | NP_065846.1 | 4 - 5 | 923 | 86 |
XM_005256744.3 | XP_005256801.1 | 3 - 4 | 873 | 86 | |
XM_017024897.1 | XP_016880386.1 | 4 - 5 | 512 | 86 | |
GenBank mRNA | AF376802.1 | - | 4 - 5 | 923 | 86 |