The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204744.1 | NP_001191673.1 | 3 - 4 | 324 | 156 |
NM_001204745.1 | NP_001191674.1 | 3 - 4 | 324 | 156 | |
NM_001204746.1 | NP_001191675.1 | 3 - 4 | 324 | 156 | |
NM_004062.3 | NP_004053.1 | 3 - 4 | 324 | 156 | |
XM_005255770.2 | XP_005255827.1 | 3 - 4 | 192 | 156 | |
XM_011522807.1 | XP_011521109.1 | 3 - 4 | 194 | 156 | |
GenBank mRNA | AF016272.1 | - | 3 - 4 | 167 | 156 |
AK075243.1 | - | 3 - 4 | 225 | 156 | |
AK225544.1 | - | 3 - 4 | 188 | 156 | |
AK225552.1 | - | 3 - 4 | 192 | 156 | |
AK298255.1 | - | 3 - 4 | 189 | 156 | |
AK298334.1 | - | 3 - 4 | 192 | 156 | |
AK313101.1 | - | 3 - 4 | 194 | 156 | |
AY358911.1 | - | 3 - 4 | 257 | 156 | |
BC027912.2 | - | 2 - 3 | 143 | 156 | |
BI759247.1 | - | 3 - 4 | 203 | 156 | |
DC361453.1 | - | 3 - 4 | 324 | 156 |