The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199699.1 | NP_001186628.1 | 9 - 10 | 1769 | 95 |
NM_020162.3 | NP_064547.2 | 10 - 11 | 1930 | 95 | |
XM_017024877.1 | XP_016880366.1 | 6 - 7 | 806 | 95 | |
GenBank mRNA | AK025625.1 | - | 10 - 11 | 1731 | 95 |
AK026944.1 | - | 4 - 5 | 447 | 95 | |
AK295074.1 | - | 9 - 10 | 1538 | 95 | |
AK295760.1 | - | 10 - 11 | 1930 | 95 | |
AK314580.1 | - | 10 - 11 | 1898 | 95 | |
AL359945.1 | - | 6 - 7 | 908 | 95 | |
BC030017.1 | - | 9 - 10 | 1456 | 95 | |
BC042040.1 | - | 7 - 8 | 1301 | 95 | |
BC050409.1 | - | 6 - 7 | 1114 | 95 | |
CR936655.1 | - | 6 - 7 | 908 | 95 |