The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173545.2 | NP_775816.1 | 6 - 7 | 977 | 145 |
GenBank mRNA | AK290841.1 | - | 6 - 7 | 977 | 145 |
AK308186.1 | - | 5 - 6 | 867 | 145 | |
BC041144.1 | - | 6 - 7 | 926 | 145 |