Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TUFT1
Entrez Gene ID
7286
Gene Name
tuftelin 1
Gene Aliases
-
Chromosome Location
Chr.1: 151540294 - 151583583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126337.1 NP_001119809.1 5 - 6 466 72
NM_001301317.1 NP_001288246.1 7 - 8 676 72
NM_020127.2 NP_064512.1 6 - 7 541 72
XM_011509961.2 XP_011508263.1 4 - 5 375 72
XM_017002223.1 XP_016857712.1 6 - 7 617 72
XM_017002224.1 XP_016857713.1 5 - 6 738 72
GenBank mRNA AF254260.1 - 6 - 7 541 72
AK075001.1 - 5 - 6 463 72
AK223161.1 - 6 - 7 537 72
AK298066.1 - 7 - 8 676 72
AK299925.1 - 5 - 6 454 72
AK315414.1 - 6 - 7 537 72
AL136917.1 - 6 - 7 529 72
AY358610.1 - 5 - 6 460 72
BC002933.1 - 5 - 6 434 72
BC008301.1 - 6 - 7 521 72

View Full Product Details