The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001312673.1 | NP_001299602.1 | 3 - 4 | 366 | 157 |
NM_005017.3 | NP_005008.2 | 4 - 5 | 430 | 157 | |
GenBank mRNA | AK309073.1 | - | 3 - 4 | 366 | 157 |
AK309300.1 | - | 3 - 4 | 371 | 157 | |
BC046355.1 | - | 4 - 5 | 391 | 157 | |
L28957.1 | - | 3 - 4 | 263 | 157 |