The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001312673.1 | NP_001299602.1 | 1 - 2 | 136 | 84 |
GenBank mRNA | AK304534.1 | - | 1 - 2 | 110 | 84 |
AK309073.1 | - | 1 - 2 | 136 | 84 | |
AK313450.1 | - | 1 - 2 | 58 | 84 |