Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HCK
Entrez Gene ID
3055
Gene Name
HCK proto-oncogene, Src family tyrosine kinase
Gene Aliases
JTK9, p59Hck, p61Hck
Chromosome Location
Chr.20: 32052188 - 32101856 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172129.1 NP_001165600.1 10 - 11 1331 76
NM_001172130.1 NP_001165601.1 10 - 11 1328 76
NM_001172131.1 NP_001165602.1 10 - 11 1328 76
NM_001172132.1 NP_001165603.1 11 - 12 1397 76
NM_001172133.1 NP_001165604.1 11 - 12 1395 76
NM_002110.3 NP_002101.2 10 - 11 1331 76
GenBank mRNA AK026432.1 - 10 - 11 1265 76
AK225819.1 - 10 - 11 1277 76
AK289896.1 - 10 - 11 1319 76
AK290928.1 - 11 - 12 1343 76
AK298726.1 - 11 - 12 1341 76
AK314335.1 - 10 - 11 1244 76
BC014435.1 - 10 - 11 1217 76
BC094847.1 - 10 - 11 1228 76
BC108930.1 - 10 - 11 1258 76
BC108931.1 - 10 - 11 1258 76
BC114463.1 - 10 - 11 1258 76
M16592.1 - 9 - 10 1106 76

View Full Product Details