The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004815.3 | NP_004806.3 | 14 - 15 | 1944 | 128 |
XM_006711048.3 | XP_006711111.1 | 13 - 14 | 1578 | 128 | |
XM_011542438.2 | XP_011540740.1 | 14 - 15 | 1819 | 128 | |
XM_011542439.2 | XP_011540741.1 | 14 - 15 | 1664 | 128 | |
XM_011542440.2 | XP_011540742.1 | 13 - 14 | 1538 | 128 | |
GenBank mRNA | AB208873.1 | - | 13 - 14 | 1550 | 128 |
AK307733.1 | - | 14 - 15 | 1773 | 128 | |
AK310491.1 | - | 14 - 15 | 1664 | 128 | |
AL833445.1 | - | 2 - 3 | 6153 | 128 | |
BC093741.1 | - | 13 - 14 | 1617 | 128 | |
BC093767.1 | - | 13 - 14 | 1620 | 128 | |
U90920.1 | - | 14 - 15 | 1758 | 128 |