Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TGM1
Entrez Gene ID
7051
Gene Name
transglutaminase 1
Gene Aliases
ARCI1, ICR2, KTG, LI, LI1, TGASE, TGK
Chromosome Location
Chr.14: 24249114 - 24263210 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000359.2 NP_000350.1 3 - 4 630 99
GenBank mRNA AK291350.1 - 3 - 4 597 99
AK299436.1 - 3 - 4 399 99
AK315819.1 - 3 - 4 630 99
AK315828.1 - 3 - 4 590 99
AK315843.1 - 3 - 4 597 99
BC034699.1 - 3 - 4 581 99
D90287.1 - 3 - 4 586 99
M55183.1 - 3 - 4 559 99
M62925.1 - 3 - 4 592 99

View Full Product Details