The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000359.2 | NP_000350.1 | 4 - 5 | 879 | 91 |
GenBank mRNA | AK291350.1 | - | 4 - 5 | 846 | 91 |
AK299436.1 | - | 4 - 5 | 648 | 91 | |
AK315819.1 | - | 4 - 5 | 879 | 91 | |
AK315828.1 | - | 4 - 5 | 839 | 91 | |
AK315843.1 | - | 4 - 5 | 846 | 91 | |
BC034699.1 | - | 4 - 5 | 830 | 91 | |
D90287.1 | - | 4 - 5 | 835 | 91 | |
M55183.1 | - | 4 - 5 | 808 | 91 | |
M62925.1 | - | 4 - 5 | 841 | 91 |