The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000359.2 | NP_000350.1 | 8 - 9 | 1421 | 58 |
GenBank mRNA | AK299436.1 | - | 8 - 9 | 1190 | 58 |
AK315819.1 | - | 8 - 9 | 1421 | 58 | |
AK315828.1 | - | 8 - 9 | 1381 | 58 | |
AK315843.1 | - | 8 - 9 | 1388 | 58 | |
BC034699.1 | - | 8 - 9 | 1372 | 58 | |
D90287.1 | - | 8 - 9 | 1377 | 58 | |
M55183.1 | - | 8 - 9 | 1350 | 58 | |
M62925.1 | - | 8 - 9 | 1383 | 58 | |
X57974.1 | - | 2 - 3 | 175 | 58 |