The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171713.1 | NP_001165184.1 | 13 - 14 | 1027 | 57 |
NM_012157.3 | NP_036289.3 | 14 - 15 | 1234 | 57 | |
XM_005265014.2 | XP_005265071.1 | 14 - 15 | 1347 | 57 | |
XM_005265015.3 | XP_005265072.1 | 14 - 15 | 1567 | 57 | |
XM_005265016.2 | XP_005265073.1 | 14 - 15 | 1311 | 57 | |
XM_005265017.3 | XP_005265074.1 | 14 - 15 | 1250 | 57 | |
XM_011533559.2 | XP_011531861.1 | 15 - 16 | 1512 | 57 | |
XM_017006092.1 | XP_016861581.1 | 15 - 16 | 1404 | 57 | |
XM_017006093.1 | XP_016861582.1 | 14 - 15 | 1394 | 57 | |
XM_017006094.1 | XP_016861583.1 | 15 - 16 | 1334 | 57 | |
GenBank mRNA | AF174589.1 | - | 15 - 16 | 1205 | 57 |
AF176518.1 | - | 14 - 15 | 1169 | 57 | |
AF186273.1 | - | 14 - 15 | 1184 | 57 | |
AK001271.1 | - | 14 - 15 | 1234 | 57 | |
AK001438.1 | - | 14 - 15 | 1202 | 57 | |
AK096492.1 | - | 1 - 2 | 1649 | 57 | |
AK294209.1 | - | 11 - 12 | 985 | 57 | |
AK297534.1 | - | 14 - 15 | 1218 | 57 | |
AK298967.1 | - | 13 - 14 | 996 | 57 | |
AK299665.1 | - | 15 - 16 | 1410 | 57 | |
AK302218.1 | - | 16 - 17 | 1462 | 57 | |
AK303764.1 | - | 12 - 13 | 1094 | 57 | |
AK316229.1 | - | 14 - 15 | 1255 | 57 | |
AK316383.1 | - | 15 - 16 | 1374 | 57 | |
AL049953.1 | - | 7 - 8 | 644 | 57 | |
BC031556.1 | - | 14 - 15 | 1178 | 57 | |
CR457121.1 | - | 14 - 15 | 1163 | 57 |