The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171713.1 | NP_001165184.1 | 3 - 4 | 186 | 71 |
NM_012157.3 | NP_036289.3 | 3 - 4 | 189 | 71 | |
XM_005265014.2 | XP_005265071.1 | 3 - 4 | 209 | 71 | |
XM_005265015.3 | XP_005265072.1 | 3 - 4 | 410 | 71 | |
XM_005265016.2 | XP_005265073.1 | 3 - 4 | 154 | 71 | |
XM_005265017.3 | XP_005265074.1 | 3 - 4 | 186 | 71 | |
XM_011533559.2 | XP_011531861.1 | 4 - 5 | 355 | 71 | |
XM_017006092.1 | XP_016861581.1 | 4 - 5 | 247 | 71 | |
XM_017006093.1 | XP_016861582.1 | 3 - 4 | 237 | 71 | |
XM_017006094.1 | XP_016861583.1 | 4 - 5 | 270 | 71 | |
GenBank mRNA | AF174589.1 | - | 3 - 4 | 160 | 71 |
AF176518.1 | - | 2 - 3 | 124 | 71 | |
AF186273.1 | - | 3 - 4 | 139 | 71 | |
AK001271.1 | - | 3 - 4 | 189 | 71 | |
AK001438.1 | - | 3 - 4 | 157 | 71 | |
AK297534.1 | - | 3 - 4 | 154 | 71 | |
AK298967.1 | - | 3 - 4 | 155 | 71 | |
AK299665.1 | - | 4 - 5 | 272 | 71 | |
AK302218.1 | - | 5 - 6 | 417 | 71 | |
AK316383.1 | - | 4 - 5 | 329 | 71 | |
AU124816.1 | - | 3 - 4 | 189 | 71 | |
BC031556.1 | - | 3 - 4 | 133 | 71 | |
CR457121.1 | - | 2 - 3 | 118 | 71 | |
DA031508.1 | - | 3 - 4 | 186 | 71 |