The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198510.2 | NP_940912.1 | 12 - 13 | 3758 | 121 |
GenBank mRNA | AY358170.1 | - | 12 - 13 | 3758 | 121 |