The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318524.1 | NP_001305453.1 | 3 - 4 | 362 | 62 |
NM_001318525.1 | NP_001305454.1 | 3 - 4 | 362 | 62 | |
NM_001318526.1 | NP_001305455.1 | 3 - 4 | 427 | 62 | |
NM_001318527.1 | NP_001305456.1 | 3 - 4 | 678 | 62 | |
NM_001318528.1 | NP_001305457.1 | 3 - 4 | 339 | 62 | |
NM_001318529.1 | NP_001305458.1 | 3 - 4 | 678 | 62 | |
NM_001318530.1 | NP_001305459.1 | 3 - 4 | 339 | 62 | |
NM_001318532.1 | NP_001305461.1 | 3 - 4 | 678 | 62 | |
NM_016209.4 | NP_057293.1 | 3 - 4 | 362 | 62 | |
NR_134670.1 | 4 - 5 | 549 | 62 | ||
NR_134671.1 | 4 - 5 | 478 | 62 | ||
XM_017023270.1 | XP_016878759.1 | 3 - 4 | 409 | 62 | |
GenBank mRNA | AF089106.1 | - | 3 - 4 | 355 | 62 |
AF161524.1 | - | 3 - 4 | 317 | 62 | |
AK126779.1 | - | 3 - 4 | 321 | 62 | |
AK307253.1 | - | 4 - 5 | 478 | 62 | |
AK310805.1 | - | 3 - 4 | 633 | 62 | |
AK311885.1 | - | 3 - 4 | 331 | 62 | |
AX771012.1 | - | 3 - 4 | 355 | 62 | |
AX771013.1 | - | 3 - 4 | 317 | 62 | |
BC011369.1 | - | 3 - 4 | 309 | 62 | |
BC018024.1 | - | 3 - 4 | 316 | 62 | |
BC105809.1 | - | 3 - 4 | 328 | 62 | |
BI754306.1 | - | 4 - 5 | 491 | 62 | |
BX384775.2 | - | 3 - 4 | 633 | 62 | |
BX419849.2 | - | 3 - 4 | 307 | 62 | |
DA209193.1 | - | 3 - 4 | 339 | 62 | |
DA293122.1 | - | 3 - 4 | 362 | 62 | |
DA295904.1 | - | 3 - 4 | 298 | 62 | |
DA833474.1 | - | 3 - 4 | 301 | 62 |