Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APOB
Entrez Gene ID
338
Gene Name
apolipoprotein B
Gene Aliases
FLDB, LDLCQ4, apoB-100, apoB-48
Chromosome Location
Chr.2: 21001429 - 21044073 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000384.2 NP_000375.2 13 - 14 1964 137
GenBank mRNA BC051278.1 - 13 - 14 1844 137
HM487065.1 - 13 - 14 1836 137
J02610.1 - 13 - 14 1914 137
M12480.1 - 3 - 4 384 137
M14081.1 - 13 - 14 1836 137
M14162.1 - 13 - 14 1836 137
X04506.1 - 13 - 14 1964 137
X04714.1 - 13 - 14 1955 137

View Full Product Details