The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000384.2 | NP_000375.2 | 16 - 17 | 2565 | 103 |
GenBank mRNA | AK290844.1 | - | 16 - 17 | 2565 | 103 |
HM487065.1 | - | 16 - 17 | 2437 | 103 | |
J02610.1 | - | 16 - 17 | 2515 | 103 | |
K03175.1 | - | 2 - 3 | 102 | ||
M12480.1 | - | 6 - 7 | 985 | 103 | |
M14081.1 | - | 16 - 17 | 2437 | 103 | |
M14162.1 | - | 16 - 17 | 2437 | 103 | |
X04506.1 | - | 16 - 17 | 2565 | 103 | |
X04714.1 | - | 16 - 17 | 2556 | 103 |