The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253727.1 | NP_001240656.1 | 15 - 16 | 1690 | 84 |
NM_001253728.1 | NP_001240657.1 | 14 - 15 | 1510 | 84 | |
NM_001253729.1 | NP_001240658.1 | 13 - 14 | 1465 | 84 | |
NM_001253730.1 | NP_001240659.1 | 14 - 15 | 1531 | 84 | |
NM_001253732.1 | NP_001240661.1 | 16 - 17 | 1693 | 84 | |
NM_001253733.1 | NP_001240662.1 | 15 - 16 | 1621 | 84 | |
NM_021634.3 | NP_067647.2 | 15 - 16 | 1609 | 84 | |
NR_045579.1 | 15 - 16 | 2226 | 84 | ||
NR_045580.1 | 15 - 16 | 1662 | 84 | ||
NR_045581.1 | 15 - 16 | 1633 | 84 | ||
NR_045582.1 | 13 - 14 | 1570 | 84 | ||
NR_045583.1 | 14 - 15 | 1549 | 84 | ||
XM_011532174.1 | XP_011530476.1 | 15 - 16 | 1656 | 84 | |
XM_011532175.1 | XP_011530477.1 | 14 - 15 | 1587 | 84 | |
XM_011532176.1 | XP_011530478.1 | 14 - 15 | 1509 | 84 | |
XM_017008517.1 | XP_016864006.1 | 14 - 15 | 1584 | 84 | |
XM_017008518.1 | XP_016864007.1 | 15 - 16 | 1578 | 84 | |
XM_017008519.1 | XP_016864008.1 | 16 - 17 | 1623 | 84 | |
XM_017008520.1 | XP_016864009.1 | 15 - 16 | 1712 | 84 | |
XM_017008521.1 | XP_016864010.1 | 16 - 17 | 1542 | 84 | |
XM_017008522.1 | XP_016864011.1 | 15 - 16 | 1627 | 84 | |
XM_017008526.1 | XP_016864015.1 | 10 - 11 | 1080 | 84 | |
GenBank mRNA | AB208943.1 | - | 16 - 17 | 1431 | 84 |
AF190500.1 | - | 15 - 16 | 1346 | 84 | |
AK122647.1 | - | 17 - 18 | 1650 | 84 | |
AK294694.1 | - | 15 - 16 | 1454 | 84 | |
AK295040.1 | - | 15 - 16 | 2049 | 84 | |
AK300379.1 | - | 14 - 15 | 1405 | 84 | |
AK307889.1 | - | 14 - 15 | 1448 | 84 | |
AK312470.1 | - | 15 - 16 | 1453 | 84 | |
AY899849.1 | - | 13 - 14 | 1202 | 84 | |
AY899850.1 | - | 14 - 15 | 1247 | 84 | |
BC112142.1 | - | 15 - 16 | 1428 | 84 | |
BC113617.1 | - | 15 - 16 | 1431 | 84 | |
BC143556.1 | - | 15 - 16 | 1451 | 84 | |
BC143558.1 | - | 15 - 16 | 1484 | 84 | |
BC143559.1 | - | 14 - 15 | 1353 | 84 | |
BX647975.1 | - | 15 - 16 | 1547 | 84 | |
BX647980.1 | - | 15 - 16 | 1547 | 84 | |
BX647985.1 | - | 15 - 16 | 1576 | 84 | |
DA308373.1 | - | 1 - 2 | 227 | 84 |