The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320766.1 | NP_001307695.1 | 5 - 6 | 786 | 112 |
NM_015565.2 | NP_056380.2 | 5 - 6 | 783 | 112 | |
XM_006723987.3 | XP_006724050.1 | 5 - 6 | 781 | 112 | |
GenBank mRNA | AB018257.2 | - | 5 - 6 | 674 | 112 |
AK001915.1 | - | 5 - 6 | 730 | 112 | |
AK023499.1 | - | 4 - 5 | 417 | 112 | |
BC029371.1 | - | 5 - 6 | 690 | 112 | |
BC140790.1 | - | 5 - 6 | 696 | 112 | |
BC150284.1 | - | 5 - 6 | 674 | 112 |