Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAMDC4
Entrez Gene ID
158056
Gene Name
MAM domain containing 4
Gene Aliases
AEGP
Chromosome Location
Chr.9: 136852367 - 136860799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206920.2 NP_996803.2 11 - 12 1329 145
GenBank mRNA AL137659.1 - 10 - 11 1626 145
AY358419.1 - 11 - 12 1329 145

View Full Product Details