The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206920.2 | NP_996803.2 | 5 - 6 | 635 | 83 |
GenBank mRNA | AL137659.1 | - | 5 - 6 | 1021 | 83 |
AY358419.1 | - | 5 - 6 | 635 | 83 |