The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206920.2 | NP_996803.2 | 8 - 9 | 987 | 100 |
GenBank mRNA | AY358419.1 | - | 8 - 9 | 987 | 100 |