Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FHOD3
Entrez Gene ID
80206
Gene Name
formin homology 2 domain containing 3
Gene Aliases
FHOS2, Formactin2
Chromosome Location
Chr.18: 36297696 - 36780220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281739.2 NP_001268668.1 14 - 15 2088 85
NM_001281740.2 NP_001268669.1 18 - 19 2664 85
NM_025135.4 NP_079411.2 15 - 16 2139 85
XM_005258349.1 XP_005258406.1 16 - 17 2457 85
XM_005258354.1 XP_005258411.1 14 - 15 2007 85
XM_005258355.1 XP_005258412.1 14 - 15 2007 85
XM_011526189.1 XP_011524491.1 19 - 20 2706 85
XM_011526190.1 XP_011524492.1 19 - 20 2706 85
XM_011526192.1 XP_011524494.1 18 - 19 2631 85
XM_011526193.2 XP_011524495.1 19 - 20 2706 85
XM_011526196.1 XP_011524498.1 16 - 17 2181 85
XM_011526197.1 XP_011524499.1 18 - 19 2631 85
XM_017026006.1 XP_016881495.1 19 - 20 2706 85
XM_017026007.1 XP_016881496.1 17 - 18 2344 85
XM_017026008.1 XP_016881497.1 19 - 20 2706 85
XM_017026009.1 XP_016881498.1 15 - 16 2121 85
XM_017026010.1 XP_016881499.1 17 - 18 2350 85
GenBank mRNA AB051482.1 - 8 - 9 1280 85
AB084087.1 - 14 - 15 1948 85
AK308859.1 - 2 - 3 736 85
BC031518.1 - 11 - 12 1591 85
HM191478.1 - 18 - 19 85

View Full Product Details