Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FHOD3
Entrez Gene ID
80206
Gene Name
formin homology 2 domain containing 3
Gene Aliases
FHOS2, Formactin2
Chromosome Location
Chr.18: 36297696 - 36780220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281739.2 NP_001268668.1 17 - 18 3328 95
NM_001281740.2 NP_001268669.1 21 - 22 3904 95
NM_025135.4 NP_079411.2 18 - 19 3379 95
XM_005258349.1 XP_005258406.1 19 - 20 3697 95
XM_005258354.1 XP_005258411.1 17 - 18 3247 95
XM_005258355.1 XP_005258412.1 17 - 18 3247 95
XM_011526189.1 XP_011524491.1 22 - 23 3946 95
XM_011526190.1 XP_011524492.1 22 - 23 3946 95
XM_011526192.1 XP_011524494.1 21 - 22 3871 95
XM_011526193.2 XP_011524495.1 22 - 23 3946 95
XM_011526195.1 XP_011524497.1 21 - 22 3649 95
XM_011526196.1 XP_011524498.1 19 - 20 3421 95
XM_011526197.1 XP_011524499.1 21 - 22 3871 95
XM_017026006.1 XP_016881495.1 22 - 23 3946 95
XM_017026007.1 XP_016881496.1 20 - 21 3584 95
XM_017026008.1 XP_016881497.1 22 - 23 3946 95
XM_017026009.1 XP_016881498.1 18 - 19 3361 95
XM_017026010.1 XP_016881499.1 20 - 21 3590 95
GenBank mRNA AB051482.1 - 11 - 12 2520 95
AB084087.1 - 17 - 18 3188 95
AK025950.1 - 3 - 4 474 95
AK091899.1 - 10 - 11 1430 95
AL834480.1 - 1 - 2 166 95
BC031518.1 - 14 - 15 2831 95
BC050670.1 - 2 - 3 250 95
BC081563.1 - 3 - 4 506 95
HM191478.1 - 21 - 22 3764 95

View Full Product Details