Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FHOD3
Entrez Gene ID
80206
Gene Name
formin homology 2 domain containing 3
Gene Aliases
FHOS2, Formactin2
Chromosome Location
Chr.18: 36297696 - 36780220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005258354.1 XP_005258411.1 10 - 11 1320 67
XM_005258355.1 XP_005258412.1 10 - 11 1320 67
GenBank mRNA AB051482.1 - 4 - 5 593 67

View Full Product Details