The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281740.2 | NP_001268669.1 | 16 - 17 | 2156 | 132 |
NM_025135.4 | NP_079411.2 | 13 - 14 | 1631 | 132 | |
XM_005258349.1 | XP_005258406.1 | 14 - 15 | 1949 | 132 | |
XM_005258354.1 | XP_005258411.1 | 12 - 13 | 1499 | 132 | |
XM_005258355.1 | XP_005258412.1 | 12 - 13 | 1499 | 132 | |
XM_011526189.1 | XP_011524491.1 | 17 - 18 | 2198 | 132 | |
XM_011526190.1 | XP_011524492.1 | 17 - 18 | 2198 | 132 | |
XM_011526192.1 | XP_011524494.1 | 16 - 17 | 2123 | 132 | |
XM_011526193.2 | XP_011524495.1 | 17 - 18 | 2198 | 132 | |
XM_011526195.1 | XP_011524497.1 | 17 - 18 | 2198 | 132 | |
XM_011526196.1 | XP_011524498.1 | 14 - 15 | 1673 | 132 | |
XM_011526197.1 | XP_011524499.1 | 16 - 17 | 2123 | 132 | |
XM_017026006.1 | XP_016881495.1 | 17 - 18 | 2198 | 132 | |
XM_017026007.1 | XP_016881496.1 | 15 - 16 | 1836 | 132 | |
XM_017026008.1 | XP_016881497.1 | 17 - 18 | 2198 | 132 | |
XM_017026009.1 | XP_016881498.1 | 13 - 14 | 1613 | 132 | |
XM_017026010.1 | XP_016881499.1 | 15 - 16 | 1842 | 132 | |
GenBank mRNA | AB051482.1 | - | 6 - 7 | 772 | 132 |
BC031518.1 | - | 9 - 10 | 1083 | 132 | |
HM191478.1 | - | 16 - 17 | 2016 | 132 |