Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBAP2
Entrez Gene ID
55833
Gene Name
ubiquitin associated protein 2
Gene Aliases
UBAP-2
Chromosome Location
Chr.9: 33921693 - 34048949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018449.3 NP_060919.3 1 - 2 71 95
GenBank mRNA AK302335.1 - 1 - 2 51 95
BG723831.1 - 1 - 2 77 95

View Full Product Details