Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBAP2
Entrez Gene ID
55833
Gene Name
ubiquitin associated protein 2
Gene Aliases
UBAP-2
Chromosome Location
Chr.9: 33921693 - 34048949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282529.1 NP_001269458.1 19 - 20 2389 62
NM_001282530.1 NP_001269459.1 7 - 8 932 62
NM_018449.3 NP_060919.3 23 - 24 2703 62
GenBank mRNA AB040924.1 - 11 - 12 1505 62
AK025801.1 - 12 - 13 1593 62
AK057564.1 - 4 - 5 1258 62
AK074884.1 - 10 - 11 1062 62
AK125935.1 - 6 - 7 2187 62
AK294952.1 - 23 - 24 2621 62
AK302754.1 - 22 - 23 2571 62
AK303493.1 - 19 - 20 2327 62
BC037416.1 - 4 - 5 336 62

View Full Product Details