Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBAP2
Entrez Gene ID
55833
Gene Name
ubiquitin associated protein 2
Gene Aliases
UBAP-2
Chromosome Location
Chr.9: 33921693 - 34048949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282529.1 NP_001269458.1 21 - 22 2687 97
NM_001282530.1 NP_001269459.1 9 - 10 1230 97
NM_018449.3 NP_060919.3 25 - 26 3001 97
GenBank mRNA AB040924.1 - 13 - 14 1803 97
AF157327.1 - 11 - 12 1216 97
AK025357.1 - 6 - 7 643 97
AK025801.1 - 14 - 15 1891 97
AK026088.1 - 17 - 18 2169 97
AK074884.1 - 12 - 13 1360 97
AK125935.1 - 8 - 9 2485 97
AK299183.1 - 19 - 20 2409 97
AK302754.1 - 24 - 25 2869 97
AK303493.1 - 21 - 22 2625 97
BC007890.2 - 3 - 4 320 97
BC037416.1 - 6 - 7 634 97

View Full Product Details