The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171976.1 | NP_001165447.1 | 24 - 25 | 3481 | 94 |
NM_017675.4 | NP_060145.3 | 24 - 25 | 3295 | 94 | |
GenBank mRNA | AB047004.1 | - | 23 - 24 | 3245 | 94 |
AK000131.1 | - | 9 - 10 | 1434 | 94 | |
AK000390.1 | - | 19 - 20 | 2849 | 94 | |
AK225217.1 | - | 9 - 10 | 1434 | 94 | |
AK315055.1 | - | 24 - 25 | 3481 | 94 | |
BC130282.1 | - | 23 - 24 | 3254 | 94 |