The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002977.3 | NP_002968.1 | 10 - 11 | 1656 | 146 |
XM_005246757.2 | XP_005246814.1 | 10 - 11 | 1629 | 146 | |
XM_011511616.2 | XP_011509918.1 | 10 - 11 | 1590 | 146 | |
XM_011511617.2 | XP_011509919.1 | 10 - 11 | 1629 | 146 | |
XM_011511618.2 | XP_011509920.1 | 10 - 11 | 1629 | 146 | |
XM_011511619.2 | XP_011509921.1 | 10 - 11 | 1629 | 146 | |
XM_017004668.1 | XP_016860157.1 | 9 - 10 | 1158 | 146 | |
XM_017004669.1 | XP_016860158.1 | 5 - 6 | 716 | 146 | |
GenBank mRNA | AK307549.1 | - | 10 - 11 | 1629 | 146 |
AY682084.1 | - | 7 - 8 | 910 | 146 | |
AY682085.1 | - | 7 - 8 | 910 | 146 | |
AY682086.1 | - | 7 - 8 | 910 | 146 | |
DQ857292.1 | - | 9 - 10 | 1356 | 146 | |
X82835.1 | - | 9 - 10 | 1363 | 146 |