Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCN9A
Entrez Gene ID
6335
Gene Name
sodium voltage-gated channel alpha subunit 9
Gene Aliases
ETHA, FEB3B, GEFSP7, HSAN2D, NE-NA, NENA, Nav1.7, PN1, SFNP
Chromosome Location
Chr.2: 166195185 - 166375987 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002977.3 NP_002968.1 22 - 23 4510 98
XM_005246757.2 XP_005246814.1 22 - 23 4516 98
XM_011511616.2 XP_011509918.1 22 - 23 4477 98
XM_011511617.2 XP_011509919.1 22 - 23 4516 98
XM_011511618.2 XP_011509920.1 22 - 23 4483 98
XM_011511619.2 XP_011509921.1 22 - 23 4516 98
XM_017004668.1 XP_016860157.1 21 - 22 4045 98
XM_017004669.1 XP_016860158.1 17 - 18 3603 98
GenBank mRNA DQ857292.1 - 21 - 22 4210 98
X82835.1 - 21 - 22 4217 98

View Full Product Details