The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022455.4 | NP_071900.2 | 2 - 3 | 1064 | 128 |
NM_172349.2 | NP_758859.1 | 3 - 4 | 279 | 128 | |
GenBank mRNA | AF380302.1 | - | 3 - 4 | 220 | 128 |
AF395588.1 | - | 2 - 3 | 971 | 128 | |
AK126591.1 | - | 2 - 3 | 1073 | 128 | |
AY049721.1 | - | 1 - 2 | 944 | 128 | |
BC139789.1 | - | 2 - 3 | 149 | 128 | |
BC150628.1 | - | 2 - 3 | 154 | 128 | |
CN333750.1 | - | 3 - 4 | 281 | 128 |