The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022455.4 | NP_071900.2 | 7 - 8 | 4330 | 115 |
NM_172349.2 | NP_758859.1 | 8 - 9 | 3545 | 115 | |
GenBank mRNA | AF380302.1 | - | 8 - 9 | 3486 | 115 |
AF395588.1 | - | 7 - 8 | 4237 | 115 | |
AK055187.1 | - | 3 - 4 | 1122 | 115 | |
AK091358.1 | - | 3 - 4 | 2137 | 115 | |
AY049721.1 | - | 6 - 7 | 4210 | 115 | |
BC144629.1 | - | 8 - 9 | 3775 | 115 | |
BC150628.1 | - | 7 - 8 | 3420 | 115 |