The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013283.4 | NP_037415.1 | 1 - 2 | 221 | 114 |
GenBank mRNA | AB073390.1 | - | 1 - 2 | 135 | 114 |
AK096178.1 | - | 1 - 2 | 123 | 114 | |
AK312365.1 | - | 1 - 2 | 170 | 114 | |
AL136664.1 | - | 1 - 2 | 134 | 114 | |
AU279489.1 | - | 1 - 2 | 170 | 114 | |
BC066645.1 | - | 1 - 2 | 96 | 114 | |
BC093030.1 | - | 1 - 2 | 130 | 114 |