The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006051.3 | NP_006042.3 | 2 - 3 | 571 | 64 |
NM_133172.2 | NP_573418.2 | 2 - 3 | 571 | 64 | |
NM_133173.2 | NP_573419.2 | 2 - 3 | 571 | 64 | |
NM_133174.2 | NP_573420.2 | 2 - 3 | 571 | 64 | |
GenBank mRNA | AB018247.1 | - | 2 - 3 | 263 | 64 |
AB049618.1 | - | 2 - 3 | 299 | 64 | |
AB061224.1 | - | 2 - 3 | 212 | 64 | |
AB079687.1 | - | 2 - 3 | 212 | 64 | |
AB094990.1 | - | 2 - 3 | 212 | 64 | |
AB209839.1 | - | 2 - 3 | 299 | 64 | |
AF224708.1 | - | 2 - 3 | 232 | 64 | |
AF224709.1 | - | 2 - 3 | 232 | 64 | |
AF224710.1 | - | 2 - 3 | 232 | 64 | |
AF224711.1 | - | 2 - 3 | 232 | 64 | |
AK075280.1 | - | 2 - 3 | 534 | 64 | |
AK125244.1 | - | 1 - 2 | 832 | 64 | |
AK307170.1 | - | 2 - 3 | 534 | 64 | |
BC013158.1 | - | 2 - 3 | 571 | 64 | |
BC125187.1 | - | 2 - 3 | 563 | 64 |