The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133174.2 | NP_573420.2 | 6 - 7 | 983 | 125 |
GenBank mRNA | AB094990.1 | - | 6 - 7 | 624 | 125 |
AF224710.1 | - | 6 - 7 | 644 | 125 |