Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HDX
Entrez Gene ID
139324
Gene Name
highly divergent homeobox
Gene Aliases
CXorf43, D030011N01Rik
Chromosome Location
Chr.X: 84317874 - 84502578 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177478.1 NP_001170949.1 6 - 7 1756 119
NM_001177479.1 NP_001170950.1 7 - 8 1903 119
NM_144657.4 NP_653258.2 6 - 7 1794 119
XM_006724619.1 XP_006724682.1 5 - 6 1647 119
XM_011530863.2 XP_011529165.1 7 - 8 1976 119
XM_011530864.1 XP_011529166.1 7 - 8 1883 119
XM_017029274.1 XP_016884763.1 6 - 7 1774 119
XM_017029275.1 XP_016884764.1 5 - 6 1720 119
GenBank mRNA AK055240.1 - 6 - 7 1730 119
AK290050.1 - 6 - 7 1768 119
BC101720.1 - 6 - 7 1706 119
BC101722.1 - 6 - 7 1706 119
BC143522.1 - 7 - 8 1815 119
BX538112.1 - 6 - 7 1766 119
BX649166.1 - 6 - 7 1769 119

View Full Product Details