Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HDX
Entrez Gene ID
139324
Gene Name
highly divergent homeobox
Gene Aliases
CXorf43, D030011N01Rik
Chromosome Location
Chr.X: 84317874 - 84502578 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177478.1 NP_001170949.1 8 - 9 1931 158
NM_001177479.1 NP_001170950.1 9 - 10 2078 158
NM_144657.4 NP_653258.2 8 - 9 1969 158
XM_006724619.1 XP_006724682.1 7 - 8 1822 158
XM_011530863.2 XP_011529165.1 9 - 10 2151 158
XM_011530864.1 XP_011529166.1 9 - 10 2058 158
XM_017029274.1 XP_016884763.1 8 - 9 1949 158
XM_017029275.1 XP_016884764.1 7 - 8 1895 158
GenBank mRNA AK055240.1 - 8 - 9 1905 158
AK290050.1 - 8 - 9 1943 158
BC101720.1 - 8 - 9 1881 158
BC101722.1 - 8 - 9 1881 158
BC143522.1 - 9 - 10 1990 158
BX538112.1 - 8 - 9 1941 158
BX649166.1 - 8 - 9 1944 158

View Full Product Details