Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HDX
Entrez Gene ID
139324
Gene Name
highly divergent homeobox
Gene Aliases
CXorf43, D030011N01Rik
Chromosome Location
Chr.X: 84317874 - 84502578 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177479.1 NP_001170950.1 3 - 4 399 92
NM_144657.4 NP_653258.2 2 - 3 290 92
XM_011530863.2 XP_011529165.1 3 - 4 472 92
XM_011530864.1 XP_011529166.1 3 - 4 379 92
XM_017029274.1 XP_016884763.1 2 - 3 270 92
XM_017029275.1 XP_016884764.1 2 - 3 270 92
GenBank mRNA AK290050.1 - 2 - 3 264 92
BC101720.1 - 2 - 3 202 92
BC101722.1 - 2 - 3 202 92
BC143522.1 - 3 - 4 311 92
BX538112.1 - 2 - 3 262 92
BX649166.1 - 2 - 3 265 92

View Full Product Details